Learn More
Thermo Scientific™ Orion™ AQUAfast™ II Chemistries Test Kits
Choose the reagent tablets you need for our colorimeters and spectrophotometers with these chemistries test kits.
Merk: Thermo Scientific™ AC2016
Bekijk meerdere types van dit product via de
Beschrijving
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
- For use with: Orion AQUAfast AQ4000, Orion AQUAfast AQ3070, Orion AQUAfast AQ3700 and discontinued Orion AQ2 series meters and othercolorimeters
- No. of tests: 50 to 100
Specificatie
Sulfide | |
Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 Colorimeter, Orion AQUAfast | |
Set of 2: 100 Sulfide No 1 Tablets, 100 Sulfide No 2 | |
Colorimetric water and wastewater testing equipment | |
0.04 to 0.5 mg/L S |
Tablet | |
AQUAfast sulfide reagent tablets, 100 tests | |
DPD/Catalyst | |
AQUAfast 24mm round vials (Cat. No. AC2V24) | |
100 Tests |
Veiligheid en behandeling
- Gevaar
Gevarencategorie
- Giftigheid voor de voortplanting BCategorie 1A
- Ernstig oogletsel/oogirritatie Categorie 1
Gevarenaanduiding
- H318-Veroorzaakt ernstig oogletsel.
- H360-Kan de vruchtbaarheid of het ongeboren kind schaden.
Veiligheidsmaatregel
- P280-Beschermende handschoenen/beschermende kleding/oogbescherming/gelaatsbescherming dragen.
- P201-Alvorens te gebruiken de speciale aanwijzingen raadplegen.
- P305+P351+P338-BIJ CONTACT MET DE OGEN: voorzichtig afspoelen met water gedurende een aantal minuten; contactlenzen verwijderen, indien mogelijk; blijven spoelen.
- P308-NA (mogelijke) blootstelling:.
- P310-Onmiddellijk een ANTIGIFCENTRUM/arts/ raadplegen.
- P405-Achter slot bewaren.
Aanvullende informatie
- MIXTURE LIST-Bevat: sodium bisulphate, boric acid, adipic acid, potassium chromate, potassium dichromate
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.