Learn More
Kaliumnitraat, 10%
Use in the outer chamber of double junction pH electrodes when using media that contains copper, lead, silver or mercury ions. Potassium Nitrate Solution, 10%, Reagecon™ is a standard solution of KNO3. It is also used as a standard in the testing of dairy products.
Merk: Reagecon EFSKNO
Beschrijving
The glass layer and reference junctions of pH electrodes need to be kept moist when not in use to remain operable and functioning at optimal levels. Potassium Chloride cannot be used with media that contains Hg++, Cu++, Ag+ or Pb++ ions, as they form insoluble precipitates with Cl– that will block the diaphragm. Potassium nitrate is a double junction bridge solution that can be used in this case.Specificatie
Colorless | |
Double junction pH electrodes | |
KNO3 | |
MFCD00011409 | |
FGIUAXJPYTZDNR-UHFFFAOYSA-N | |
potassium nitrate | |
Miscible in Water | |
Potassium Nitrate |
10% w/v | |
101.102 | |
100 ml | |
7757-79-1 | |
[K+].[O-][N+]([O-])=O | |
101.10 | |
Liquid |
Veiligheid en behandeling
- ELECTRODE FILLING SOLUTION 10% POTASSIUM NITRATE
Signaalwoord
- Gevaar
Gevarencategorie
- Oxiderende vloeistof Categorie 2
Gevarenaanduiding
- H272-Kan brand bevorderen; oxiderend.
Veiligheidsmaatregel
- P210-Verwijderd houden van warmte, hete oppervlakken, vonken, open vuur en andere ontstekingsbronnen. Niet roken.
- P220-Van kleding/brandbare stoffen verwijderd houden/bewaren.
- P280-Beschermende handschoenen/beschermende kleding/oogbescherming/gelaatsbescherming dragen.
- P370+P378-In geval van brand: blussen met .
- P501-Inhoud/verpakking afvoeren naar .
Aanvullende informatie
- MIXTURE LIST-Bevat: potassium nitrate