Learn More
Sartorius Sartolab™ P20-filtersystemen
Filter weefselcultuuroplossingen snel, economisch en betrouwbaar. Sartorius™ Sartolab™ P20 Plus-filtersystemen zijn steriele, gebruiksklare eenheden. Een groot filtratieoppervlak en automatische ontluchting zorgen voor hoge stroomsnelheden. Verkrijgbaar met PES- of SFCA-membraan, verschillende connectoren of met een prefilter voor de verwerking van maximaal 10 l.
Merk: Sartorius 18068----------D
156.99 EUR valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Beschrijving
Filter tissue culture solutions quickly, economically and reliably. Sartorius Sartolab™ P20 Plus Filter Systems are sterile, ready-to-use units. A large filtration area and automatic de-aeration ensures high flow rates. Available with PES or SFCA membrane, different connectors or with a prefilter to process up to 10L.
The new way for reliable sterile filtration- Ready-to-use unit with a large filtration area (20cm2) for fast filtration
- Quickly and reliably sterilizes media and aqueous solutions with optimal throughputs
- Automatically vents any trapped air through the PTFE membrane-protected vent ports
- Extraordinarily favorable price-performance ratio for a cost-effective solution
- The total filterable volume can often be doubled due to an integrated binder-free, glass fiber pre-filter
- High-quality construction; every batch is rigorously tested to ensure sterility and integrity
Specificatie
USP, klasse VI | |
Single-use | |
Polycarbonate | |
0.2 μm | |
Sterile | |
Betrouwbare steriele filtratie van weefselcultuuroplossingen | |
3 bar | |
10 Pack | |
100 mL to 10 L |
Luer lock / slangnippel | |
Ready-to-Use Pressure Filter | |
Polyethersulfon | |
100 mL to 5 L | |
20 cm2 | |
Glass Prefilter, Filling Bell | |
5 bar | |
Pressure Filter | |
0.9 mL |