Learn More
BD Difco™ gedehydrateerd kweekmedium: Gist op stikstofbasis zonder aminozuren en ammoniumsulfaat
Wickerham-formule, classificatie van gisten gebaseerd op stikstof- en koolstofvereisten
Merk:  BD 233520
 Bekijk meerdere types van dit product via de
Produktcode. 11743014
  • 80.20 /
Edge
Beschrijving
Beschrijving
Media with Yeast Nitrogen Base without Amino Acids and Ammonium Sulfate are included in many applications for the study of yeasts in molecular genetics. Yeasts are unicellular, eukaryotic, budding cells that are generally round-to-oval or elongate in shape. They multiply principally by the production of blastoconidia (buds). Yeast colonies are moist and creamy or glabrous to membranous in texture. Yeasts are considered opportunistic pathogens.
  • Contains all essential nutrients and vitamins necessary for the cultivation of yeasts and a source of nitrogen and carbohydrate
  • Lacks the amino acids histidine, methionine and tryptophan and ammonium sulfate
  • Prepared according to Guenter’s modification of Wickerham’s Yeast Nitrogen Base formulation
  • Displays a good cultural response to Kloeckera apiculata 9774 and Saccharomyces cerevisiae 9080
  • pH (0.17% solution, 25°C): 4.5 ± 0.2
Informatie over bestelling
Prices are for units of one each unless otherwise specified.
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
ISO/DIS, 11014
<30 °C
Gist, stikstofbasis
Vitamines: Biotine 2 μg/l, panthoteenzuur 400 μg/l, foliumzuur 2 μg/l, inositol 2000 μg/l, niacine 400 μg/l, p-aminobenzoëzuur 200μ g/l, pyridoxine
Fles
Light yellowish-beige
Gedehydrateerd
Microbiology Media
Voor classificering van gist op basis van kolonie-eigenschappen en celmorfologie
100 g
Product Suggestions
Product Suggestions
Video's
Veiligheid en behandeling
Veiligheid en behandeling
Productidentificatie
  • Yeast Nitrogen Base w/o Amino Acids & Ammoni
Aanvullende informatie
  • MIXTURE LIST-Bevat: calcium chloride