Learn More
Fisherbrand™ Met platina behandelde siliconenslangen, doorzichtig
Providing a number of advantages over conventional peroxide cured silicones. Fisherbrand™ Translucent Platinum Cured 5m Silicone Tubing is available in a comprehensive range of stock sizes.
Merk: Fisherbrand™ 1172-8118
This item is not returnable.
View return policy
Beschrijving
- Autoclavable
- Non-yellowing
- High gloss finish
- Excellent quality
- Highly hydrophobic
- No peroxide byproducts
- High tear strength as standard
- Extended life in Peristaltic pumps
- Applications include biological, pharmaceutical, biotechnology, aerospace and high temperature work
Specificatie
FDA, BFR and WRAS approved | |
4 mm | |
5.6 mm | |
0.8 mm | |
-40 tot 200 °C | |
Flexible | |
-40 tot 200 °C |
Ja | |
Platinasilicone | |
5 m | |
Ideaal voor biologische, farmaceutische en biotechnologische werkzaamheden, de ruimtevaart en brandgevaarlijke werkzaamheden | |
Doorzichtig | |
0.5 bar |