Learn More
Zwiebel™ Gewicht met COFRAC kalibratiecertificaat, klasse E2
Calibrate weights of lower accuracy classes. Zwiebel™ Class E2 Individual Stainless Steel Leaf Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
Merk:  Zwiebel™ PBI2302S
 Bekijk meerdere types van dit product via de
Produktcode. 11700643
  • 200.00 / 1 stuk
Only null left
Toevoegen aan winkelmandje
Description
Description
  • Class E2 calibration weight
  • In compliance with OIML R111
  • Delivered with a COFRAC calibration certificate guaranteeing traceability
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
100 mg
±0.016 mg
For use with Weighing Instruments of Special Accuracy Class I