Learn More
Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter
Reliably measure pH, mV, ORP and temperature with our benchtop pH meter for advanced lab analysis and printer or computer interfacing.
Merk: Thermo Scientific™ STARA2115
1021.58 EUR valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Beschrijving
Record accurate and reliable pH, mV, ORP and temperature measurements with the Thermo Scientific™ Orion Star™ A211 Benchtop pH Meter. This meter is ideal for a wide range of applications and advanced pH analysis in the lab. Perform up to a 5 point pH calibration, log up to 2000 data point sets with time/date stamp and easily transfer calibration and data logs via USB or RS232 to a printer or computer with free downloadable Orion Star™ Com software. Use the included electrode stand to easily place sensors in samples and simplify storage to minimize breakage.
Thermo Scientific Orion Star A211 Benchtop pH Meters are easy-to-use, dependable laboratory meters with advanced features and functions. Selectable read modes and stability indicator give you flexibility and control over your testing. All critical data is shown with your measurements including temperature, date and time, read type, measurement stability, active calibration information and electrode condition icon. Onscreen calibration and setup instructions can be displayed in several local languages with new languages added through software updates.
Navigation is fast and simple due to the comprehensive keypad with menu-specific function keys, dual purpose scroll/shortcut keys, plus power, measure, log view and stirrer keys that ensure easy operation. All setup menus are in one location with direct access using a single shortcut key so you can quickly access, evaluate and change measurement and instrument settings.
- Measure pH, mV, relative mV or ORP with temperature
- Perform up to a 5 point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values
- Calibration editing option allows individual points to be fixed without a full recalibration
- Informative, easy-to-read backlit graphic display with clear onscreen instructions simplifies operation
- Capture your critical readings with measurement stability indicator and selectable read modes — Auto-Read holds a stable reading, Continuous shows changing readings with hold option and Timed logs data at specific intervals
- Eliminate the need for magnetic stir bars using the meter-controlled stirrer probe (catalog number 096019) to uniformly mix solutions
- Data log collects and date/time stamps up to 2000 measurement sets with optional sample ID and user ID
- Calibration log stores 10 most recent calibrations
- Non-volatile meter memory ensures your data and settings are protected, even with loss of power
- Orion Star Com software facilitates data transfer to a computer via USB or RS232 connections
- Flexible power options with universal power adapter or optional AA batteries
- IP54-rated housing resists dust and splashes and can be wall-mounted if table space is limited
- Meters include CE, TUV 3-in-1 and FCC Class A certifications
- 3-year replacement warranty
Specificatie
Orion Star A211 kit voor pH-meter, tafelmodel, met ROSS Ultra Triode pH/ATC-elektrode met glazen behuizing, oplossingen en standaard | |
±0.1 °C | |
pH ±0.002 | |
USB, RS-232 | |
pH met mogelijkheid tot bewerken kalibratie, relatieve mV (RmV), ORP en temperatuur | |
pH 0.1, 0.01, 0.001 | |
±2000.0 mV | |
1 tot 5 | |
22.0°F to 221.0°F | |
Handmatig of automatisch met ATC-temperatuursonde | |
Grafische lcd met backlight | |
Logboekbestanden, kalibratiebestanden en meterinstellingen blijven bewaard dankzij niet-vluchtig geheugen | |
Automatische gegevensregistratie met Auto-Read en Timed meetmodi; handmatige gegevensregistratie met | |
BNC (pH- of ORP-elektrode), pinpunt (referentie-elektrode), 8-pins mini-DIN (ATC-temperatuursonde), invoer roerder (sonde met roerder) | |
IP54 | |
24 cm x 18 cm x 11 cm (9.5 inch x 7.1 inch x 4.3 inch) | |
2 lb. | |
Uitgebreid, met menuspecifieke functietoetsen en toetsen met tweeledige functie (scroll-/sneltoetsen) | |
Orion Star A211 pH-/ mV-/RmV-/ORP-/temperatuurmeter, tafelmodel | |
pH-elektrode, ORP-elektrode, referentiehalfcel-elektrode, ATC-temperatuursonde, sonde met roerder (inclusief 8302BNUMD ROSS Ultra Triode pH/ATC-elektrode) | |
±2000.0 mV | |
pH 7.000 | |
41°F to 113°F (ambient) | |
1 | |
pH Meter TRIS Kit |
Star A211 pH-meter, 8302BNUMD ROSS Ultra Triode pH/ATC-elektrode met glazen behuizing, 810199 ROSS kit voor pH-buffer en oplossing, elektrodestandaard, universele stroomadapter, documentatie-cd, papieren snelstartgids, computerinterfacekabel en certificaa | |
±0.2 mV of, als dat meer is, ±0.05% van de meetwaarde | |
Datapunten (afzonderlijke, binnen een bepaald bereik of alle) kunnen worden overgezet naar printer o | |
pH, mV, relatieve mV (RmV) of ORP met temperatuur, mV, Relative mV, ORP with Temperature | |
pH -2.000 tot 20.000 | |
±2000.0 mV | |
0.1 mV | |
-5.0 °C tot 105.0 °C, 22.0 °F tot 221.0 °F | |
0.1 °C, 0.1 °F | |
1-punts kalibratie van de temperatuurafwijking | |
Ja, selecteerbaar | |
2000 met datum- en tijdstempel | |
Waarschuwing bij bereiken boven-/onderlimiet, waarschuwing wanneer kalibratie nodig is | |
CE, TUV 3-1, FCC-klasse A | |
Universele stroomadapter, 50-60 Hz, 100-240 VAC (cat. Nr. 1010003) | |
0.9 kg | |
3 jaar garantie op de meter | |
10 per kanaal met wachtwoordbeveiliging | |
Universele AC-adapter (inbegrepen) of 4 AA-batterijen (afzonderlijk aan te schaffen) | |
Engels, Spaans, Frans, Italiaans, Duits en Chinees; Portugees en Koreaans beschikbaar via software-update | |
5 tot 85%, zonder condensatie | |
Naar keuze snel, gemiddeld, langzaam of Smart Stability-instelling; naar keuze uitgeschakeld of automatische Smart Averaging-instelling | |
5 °C tot 45 °C (omgevingstemperatuur) | |
Vier AA (afzonderlijk aan te schaffen) | |
800 uur |