Learn More
Gibco™ StemPro™-34 SFM (1X)
A serum-free medium specifically formulated to support the development of human hematopoietic cells in culture.
Merk: Gibco™ 10639011
262.20 EUR valid until 2025-12-31
Use promo code "24342" to get your promotional price.
Beschrijving
- A serum-free medium specifically formulated to support the development of human hematopoietic cells in culture
- This includes HSC and progenitor cells isolated from bone marrow, neonatal cord blood, and peripheral blood
- StemPro-34 SFM is a flexible cell culture media that can be used for a wide variety of applications
Specificatie
Hematopoietic Stem Cell, Hematopoietic Cell, Bone Marrow | |
Liquid | |
Serum-free | |
No Glutamine | |
500 mL | |
Human | |
Stationary Suspension Culture |
Serum-free | |
Stem Cell Serum Free Media (SFM) | |
Phenol Red | |
StemPro™ | |
≤ 10 EU/mL | |
• 500 ml StemPro™-34 Serum Free Medium (store at 2°C to 8°C in the dark) • 13 ml StemPro™-34 Nutrient Supplement (store at -5°C to -20°C in the dark) Store the complete medium at 2°C to 8°C in the dark. |
For human ex vivo tissue and cell culture processing applications. CAUTION: When used as a medical device, Federal Law restricts this device to sale by or on the order of a physician.