Learn More
Fisherbrand™ Buisjes van borosilicaatglas, met schroefdop van polypropyleen, voor eenmalig gebruik
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
Merk: Fisherbrand™ 73750M-102
560.50 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Beschrijving
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes
- Natural linerless polypropylene screw cap
- With marking spot
Ideal for tissue culture, bacteriology, clinical chemistry, blood typing, and crossmatching procedures
Informatie over bestelling
Verpakt in trays van 250 of 500
Specificatie
25 mL | |
Borosilicate Glass | |
Provides chemical stability and biological inertness of hard, borosilicate glass at price of disposable tubes | |
Tray | |
Non-sterile | |
Screw Cap |
125 mm | |
Single-use | |
Marking Spot | |
250/Tray | |
20 mm | |
Disposable Culture Tube |