Learn More
Gibco™ Fetal Bovine Serum, dialyzed, US origin
GREENER_CHOICE
Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.
Merk:  Gibco™ 26400044
679.38 EUR valid until 2025-06-30
Use promo code "24108" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 11520646
  • 1014.00 /
Edge
Beschrijving
Beschrijving
Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Storage conditions: ≤-10°C
Shipping conditions: Frozen
Fetal
Liquid
Fetal Bovine Serum
Dialyzed
Cattle/Bovine
United States
Bottle
500 mL
Frozen
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.