Learn More
Cytiva Benchkote™-oppervlaktebescherming
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
Merk:  Cytiva 2300-731
Produktcode. 11380664
  • 316.00 / Each
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
Minimize mess and contamination during spills. Whatman™ Benchkote material has a high-quality, smooth, absorbent lab paper layer that soaks up liquid spills. A second laminated polyethylene layer prevents flow-through to the working surface.
  • Strong: Benchkote surface protector is strong and tear resistant when wet or dry
  • Ease-of-use: Smooth white surface can be written on with ink or pencil and lies flat
  • Clean: Suitable for saturation with disinfectant to protect benches where pathogens and other bacteria are present
  • Disposable: Benchkote paper can be incinerated after use
  • Safe combustion: Polyethylene layer is rapidly consumed when incinerated and does not melt or drip
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Werkbankbeschermer - rol
0.75 l/m2
460 mm
50 m
Geschikt voor verzadiging met desinfecterend middel
Polyethyleen
18 in.
164 ft.
18 x 164 voet (460 x 50 m)
Elk