Learn More
Maxima Maxima™ SYBR™ Green 2X qPCR Master Mix met ROX™-oplossing
Ready-to-use solution optimized for qPCR and 2-step RT-qPCR.
Merk:  Maxima K0253
3056.46 EUR valid until 2025-08-24
Use promo code "24117" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 11340760
  • 3935.00 / 4000 reactions
Beschrijving
The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added. The SYBR Green I intercalating dye allows for DNA detection and analysis without using sequence-specific probes.
Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima SYBR Green qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates. Maxima SYBR Green qPCR Master Mixes are compatible with most real-time thermal cyclers.
  • Specificity - Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Sensitivity - detects low copy number targets
  • Wide linear range - accurate quantification across 9 orders of magnitude
  • Reproducibility and convenience - ready-to-use 2X master mix
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Contains:
• 4 x 12.5 mL Maxima SYBR Green qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase, dNTPs (also dUTP) and SYBR Green I in an optimized PCR buffer; sufficient for 4000 x 25 μL reactions.
• 1 mL ROX solution, 50 μM
• 2 x 30 mL Nuclease-Free Water
Store at -20°C.
Tube
qPCR
Standard
Maxima
4,000 reactions
Gene Expression
4,000 Reactions
4 x 12.5 mL
SYBR
High
Taq DNA Polymerase
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
Real Time PCR SYBR Master Mix
4000 Reactions of 25 μL
2X
DNA, dsDNA