Learn More
Bel-Art™ Plate Reader
Tool allows safe examination of the bottom of plates without risking spills on the face and eyes
Merk:  Bel-Art™ F37861-0000
Produktcode. 11328763
  • 268.00 /
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • For examining cell monolayer, cell clones and for counting viral plaques or bacterial colonies
  • Easy and safe reading of hemagglutination, hemagglutination inhibition, hemolysis and agglutination tests
  • Clear deck for resting plates
  • Large, adjustable mirror is perfect for viewing plate underside
  • Mirror magnification is 3X actual plate size
  • Made of polypropylene with a glass-clear acrylic deck for placement of plates and Petri dishes
Hemagglutination, Hemagglutination Inhibition, Hemolysis and Aglutination testing
Specificatie
Specificatie
Microtiter Plate Reader
3X
Hemagglutination, hemagglutination inhibition, hemolysis and agglutination testing
None
Polypropylene, Clear Glass Acrylic Deck
Microtiter plates