Learn More
Thermo Scientific Chemicals Methylene Blue, 1% w/v aq. soln.
CAS: 122965-43-9 | C16H18ClN3S
Merk:  Thermo Scientific Chemicals 042771.AE
31.80 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 11305597
  • 32.00 /
Beschrijving
Methylene Blue, 1% w/v aqueous solution is used as a dye in different staining procedures viz. Wright's stain and Jenner's stain. It also serves as an indicator and medicine. Further, it is used to examine RNA and DNA under the microscope or in a gel. It is widely used as a redox indicator in analytical chemistry. It is also used in the sulfide analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Methylene Blue
1% w/v aqueous solution
MFCD00012111
319.86
23 hPa (17mm Hg) at 20°C
Liquid
122965-43-9, 7732-18-5
C16H18ClN3S
Miscible with ethanol,chloroform,glacial acetic acid and glycerol. Slightly miscible with pyridine. Immiscible with ethyl ether,xylene,oleic acid,ethanol and acetone.
3, 7-Bis(dimethylamino)phenothiazin-5-ium chloride hydrate: 1.0%; Water: 99%
100 mL
RUO – Research Use Only