Learn More
Merck Steriele Millex™-spuitfilters: MCE-membraan - blauw
Ontworpen voor steriliseren en zuiveren van steriele oplossingen. Steriele EMD Millipore Millex™-spuitfilters: MCE-membraan zijn gebruiksklare spuiteenheden die tijdens de filtermontage nooit door mensenhanden zijn aangeraakt. Verkrijgbaar met poriegroottes van 0.22, 0.45 en 0.80 μm; filters van medische kwaliteit verkrijgbaar.
Merk: Merck SLAA033SS
Beschrijving
Designed for sterilizing and clarifying sterile solutions. Millex™ Sterile Syringe Filters: MCE Membrane are ready-to-use, syringe-driven units that have never been touched by human hands during filter assembly. Feature 0.22, 0.45 and 0.80μm pore sizes; medical-grade filters available.
Sterile Millex filter units are available in a larger 33 mm housing with either MCE, PVDF or PES membranes.Faster flow rate: The larger filter surface area increases flow rate and throughput. It also makes it easier to filter solutions because it reduces the pressure required to empty the syringe.
Higher operating pressure: Millex filters have a maximum housing pressure of 10.5 bar (150 psig), which means you can filter solutions faster than before.
Color-coded: The color-coded band on the Millex housing clearly indicates which membrane is inside.
Manufactured for reliable performance: Millex filters are manufactured in a controlled environment using an automated process. Human hands never touch the filter during assembly. A Certificate of Quality included in each box describes Millipore's quality standards in detail.
Choice of membranes:Millex filters are available with three membranes: Millipore Express™ (PES) membrane, mixed cellulose esters (MCE), and Durapore (PVDF).
- Millex-GP filters feature Millipore Express (PES) membrane, which provides a unique combination of speed and six times less protein binding than competitive PES membranes. Millex-GP filters are recommended for sterile filtering protein solutions, tissue culture media, buffers and additives.
- Millex filters with Durapore™ (PVDF) membrane are the lowest protein-binding syringe filters available. Use them to filter protein solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration.
- Millex-MF filters feature mixed cellulose esters (MCE) membrane, which provides reliable, general purpose filtration of water or other aqueous solutions. The 0.2 μm filters are sterilizing grade. Larger pore sizes are for clarification and prefiltration.
Applications:Sterilization of Aqueous Solutions, Organic Solvents and Gases, Filtration of High Value Protein or Small Molecule Components.
Compliance
Voldoen aan USP klasse VI voor biologische tests voor kunststoffen. Alle filters van 33 mm zijn voorzien van een CE-markering en zijn geschikt voor zowel medische als laboratoriumtoepassingen buiten de V.S.
Specificatie
Not Autoclavable | |
Blauw | |
Membrane Filter | |
0.8 μm | |
Gebruik voor sterilisatie/bezinking van water, buffers en zoutoplossingen | |
Mixed Cellulose Esters (MCE) | |
mannelijke Luer-Slip | |
50 per verpakking | |
45°C | |
45 °C | |
Injectiespuitfilter | |
33 mm |
Voldoet aan USP-klasse VI voor biologische tests voor kunststoffen. | |
Single-use | |
MCE | |
Sterile | |
<100 μL | |
Luer-Lock-connector | |
145 psi | |
113°F | |
113°F | |
Gemodificeerd acryl | |
<200 ml |