Learn More
Pyrex™ horlogeglazen van borosilicaatglas
Manufactured from heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Watch Glasses are used for evaporation work and covering heated beakers. Fire-polished edges reduce chipping and breakage.
Merk: Pyrex 1060/06D
143.45 EUR valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Manufactured from heat resistant Pyrex™ Borosilicate Glass. Pyrex™ Borosilicate Glass Watch Glasses are used for evaporation work and covering heated beakers. Fire-polished edges reduce chipping and breakage.
- For evaporation work and covering heated beakers
- Manufactured from heat resisting Pyrex™ borosilicate glass
- Fire-polished edges to reduce chipping and breakage
Specifications
Flat | |
Round | |
Voor verdampingswerkzaamheden en afdekken van verwarmde bekers | |
50 mm |
Pyrex-borosilicaatglas | |
Watch Glass | |
Polished |