Learn More
Ratiolab™ Standaard en afvalzakken, tafelmodel
Use these bags for the disposal of contaminated labware. Waste Disposable Bags and Benchtop Stand set consists of bench-top stand and distinctive red autoclavable bags for easy identification.
Merk:  Ratiolab™ 7001401
Produktcode. 10631246
  • 31.10 /
Only null left
Edge
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • 100 red autoclavable Bags with BIOHAZARD warning Instruction and sterilization indicator patch
  • Bags measure 210 x 290mm, made of robust polyethylene (PE)
  • Rectangle white indicator patch field turns into dark when autoclaved
  • With BIOHAZARD symbol and safety imprint in English, German, French and Spanish languages
  • Autoclavable at 121 °C, for autoclaving, the bag must not be completely closed
  • Practical bench-top stand, plastic-coated, with 3 rubber feet
Specificatie
Specificatie
Set met tafelstandaard en afvalzakken
2.5 l/zak
100 afvalzakken
200 x 300 mm