Learn More
Falcon™ Falcon™-buisjes met ronde bodem, polystyreen
Gemaakt van optisch helder polystyreen voor een goede observatie van het monster. Corning™ Falcon™-buisjes met ronde bodem van polystyreen worden voor veel toepassingen gebruikt en zijn vaak beschreven in laboratoriumprotocollen.
Merk: Falcon™ 352052
129.58 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Beschrijving
- Widely used and referenced in laboratory protocols
- Sterile, nonpyrogenicity tested to less than 0.1 EU/mL (Cat. No. 352008 nonsterile)
The foundation for consistent research results
- Provide reliable containment of laboratory fluid samples
- Polystyrene tubes are best suited for procedures requiring high optical clarity
- Medical-style packaging materials
- 1,400 RCF rating (RCF claims refer to Relative Centrifugal Force measured in g-force for materials with a specific gravity of 1.0, used in an appropriate rotor with correct cushion and safety precautions. Tubes used with organic solvents at temperatures below 0°C may have lower RCF ratings.)
- Tube dimensions and volumes are approximate
Specificatie
Transparant | |
5 ml | |
Polystyreen | |
1000/Cs. | |
No | |
nr.; Zonder |
1400 x G | |
75 mm | |
Ja | |
Sterile | |
12 mm | |
Reageerbuisje |