Learn More
Macherey-Nagel™ Glasplaten, silica
Gescheiden niet-vluchtige mengsels met ongemodificeerde standaard silicalagen voor TLC (dunnelaagchromatografie). Standaard SIL G silicalagen op glazen platen zijn verkrijgbaar in verschillende plaatafmetingen en laagdikten. Selecteer een fluorescentie-indicator en kies uit de verschillende verpakkingsformaten.
Merk:  Macherey-Nagel™ 809027
141.78 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 10575571
  • 159.12 /
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G silica layers on glass plates come in a variety of plate sizes and layer thicknesses. Select a fluorescent indicator and choose from a variety of pack sizes.
Glass TLC plates, SIL G/UV 254.
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specificatie
Specificatie
Silica 60
60 Å
Glasplaten
0.75 ml/g
0.25 mm
5 - 17μ m
50 per pak
UV 254
∽ 500 m2/g
50 x 100 mm