Learn More
Greiner Bio-One Cryogeen buisje van polypropyleen, met inwendige schroefdraad
Manufactured with high-grade polypropylene for strength and high thermal resistance. Greiner BioOne™ Cryo.s Cryogenic Tubes with Internal Threads withstand a temperature range of -196 to +121°C.
Merk: Greiner Bio-One 121278
This item is not returnable.
View return policy
Beschrijving
- Free of detectable human DNA, endotoxins, DNase and RNase
- Sterile, non-pyrogenic, non-cytotoxic, no heavy metals contained
- CE-marked (CE/IVD)
- As a safety precaution Cryo.s should be stored in a freezer or exclusively in the gas phase over the liquid nitrogen
- Suitable for shipment by air
- With white, scratch-resistant writing area
- 150 inserts per case (for additional labelling on the cap)
- Suitable for storage at -20°C, -80°C and in the vapour phase of LN2
- Multiple cap colours and working volumes
- Ultra-pure medical grade, USP class VI certified polypropylene w/o leachables
- Accessory storages boxes and working racks available
Specificatie
Cryogenic Storage Tube | |
Not Autoclavable | |
Round (Self-standing) | |
Writing Area, Cap Inserts | |
Sterile | |
Polypropylene |
Yellow | |
2 mL | |
Polypropylene | |
-196°C to +121°C | |
Yes | |
48 mm |