Learn More
Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes
BIOPHARMAGREENER_CHOICE
Store samples from general cold storage to the vapor phase of liquid nitrogen with these biobanking and cell culture cryogenic tubes, available with either internal or external threads.
Merk:  Thermo Scientific™ 375353
250.43 EUR valid until 2025-12-31
Use promo code "24325" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 10544641
  • 398.00 /
Edge
Beschrijving
Beschrijving
Store samples from general cold storage to the vapor phase of liquid nitrogen with Thermo Scientific™ Nunc™ Biobanking and Cell Culture Cryogenic Tubes, available with either internal or external threads.
  • Non-cytotoxic, non-mutatgenic raw materials and pyrogen-free IVD compatible tubes for added sample security
  • External or internally threaded options available
  • Certified SAL of 10-6 for the highest sterility assurance available
  • Suitable for storage from general cold storage refrigeration (+4°C) down to the vapor phase of liquid nitrogen
  • Bulk, uncapped, non-sterile versions available for less demanding applications
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
12.5 mm
With writing surface
Not Autoclavable
Conisch
Extern
41 mm
Losstaand
Ja
Steriel
500/Pk.
0.0338 oz.
Screw Cap
Cryogenic Storage Tube
0.492 in.
1 ml
Polypropylene
1.614 in.
500
Ja
Nunc™
Ja
Polyethyleen (PE)