Learn More
Carl Roth™ Universeel absorbens
Ideal for absorption of inflammable, oily, and aggressive liquids. Roth Carl™ Universal Absorber Rotisorb™ makes working with volatile liquids safer.
Merk: Carl Roth™ 1710.1
Beschrijving
Ideal for absorption of inflammable, oily, and aggressive liquids such as:
- anorganic and organic acids
- alkaline solutions
- alcohols
- aldehydes
- aminesesters
- aromatic, chlorinated and aliphatic hydrocarbons
- substance compounds, e. g. paint, heating oil, fuels, lacquers, motor lubricants
Specificatie
Universeel absorbens | |
1 per pak |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.