Learn More
Lenz Laborglasinstrumente™ Conische scheitrechters van borosilicaatglas met glazen stop
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Separating Funnels, with Glass Plug, without Scale are manufactured from durable DURAN™ borosilicate glass.
Merk:  Lenz Laborglasinstrumente™ 04000070
 Bekijk meerdere types van dit product via de
Produktcode. 10465601
  • 133.00 / 2 stuks
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Without scale; with solid glass plug
  • With NS socket and PE stopper
  • With screwthread retaining device
  • Corrosion-free and resistant to thermal shock
Specificatie
Specificatie
1000 ml
Borosilicaatglas
Pakket 2
Polyethyleen