Learn More
Bochem™ 18/10 roestvrijstalen chemische lepel
Stainless steel spoon for scooping chemicals.
Merk: Bochem™ 3423
This item is not returnable.
View return policy
Beschrijving
- Chemical spoon
- With spatula handle
Specificatie
Elk 1 | |
32 mm | |
1 mm | |
210 mm |
Chemische lepel | |
22 mm | |
18/10 roestvrij staal |