Learn More
Merck Express™ PLUS-membraanfilters
Plain white polyethersulfone Express™ PLUS Membrane Filters provide ultrafast filtration for a variety of applications.
Merk: Merck GPWP04700
Beschrijving
Plain white polyethersulfone membranes provide ultrafast filtration for a variety of applications
The Millipore Express™ PLUS membrane provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
Features and Benefit:- Ultra fast filtration and high throughput shorten process time
- Can be sterilized by autoclave, ethylene oxide, or gamma irradiation for sterile filtration applications
- Applications:Sterile Filtration, Buffer Filtration, Tissue Culture Media Filtration
Specificatie
Ja | |
Reusable | |
Disc | |
0.2 μm | |
15 (at 690 Mbar (10 psi)) | |
Hydrofiel | |
Filter Membranes | |
1.9 in. |
Wit | |
Screen Filter | |
Polyethersulfon (PES) | |
Non-sterile | |
Voor het extra snel filtreren van weefselcultuurmedia, additieven, buffers en andere waterige oplossingen. Dit membraan wordt ook gebruikt in vele gebruiksklare steriele filtratieapparatuur. | |
Plain | |
Hydrophilic | |
47 mm |