Learn More
Buerkle™ twee- en driewegkleppen
Gebruik deze kleppen voor de overdracht van vloeistoffen of gassen om de stroomrichting vrij te geven, te blokkeren of te veranderen. Buerkle™-twee- en driewegskleppen zijn verkrijgbaar in PVDF- en PP/PE-constructie voor diverse toepassingen.
Merk: Buerkle™ 8606-0100
15.45 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Beschrijving
For transfer of liquids or gases in order to release the flow, to block it or to change its direction
PVDF Three Way Valves:
- excellent chemical resistance, transparent, suitable for food
- Operating temperature: +0 up to +40 °Resistant to compression max. 1 bar
- With mounting plate for easy installation
PP/PE Three Way Valves:
- for industrial use, red/white
- Operating temperature: +5 up to +40 °C
- Resistant to compression max. 1 bar
- With mounting plate for easy installation
Specificatie
Ja | |
Rood/wit | |
Mounting Plate | |
Polypropylene, Polyethylene | |
8 mm | |
8 mm |
2011/65/EC | |
9 to 11 mm dia. Hose | |
40°C | |
5°C | |
3-way Valve |