Learn More
Thermo Scientific™ Oxoid™ tabletten voor fosfaatgebufferde zoutoplossing
Oxoid Phosphate Buffered Saline Tablets (Dulbecco A) is used for preparation of balanced salt solution without calcium and magnesium.
Merk: Thermo Scientific™ BR0014G
55.64 EUR valid until 2025-12-31
Use promo code "24336" to get your promotional price.
Beschrijving
Oxoid Phosphate Buffered Saline Tablets (Dulbecco A) is used for preparation of balanced salt solution without calcium and magnesium.
Specificatie
Tabletten met fosfaatgebufferde zoutoplossing | |
Media Supplement | |
Elk tablet voor 100 mL |
Tablet | |
Elk |
For Laboratory Use Only