Learn More
Nabertherm MULTITHERM OVEN 15L 650°C PID
Merk:  Nabertherm 001334160
MULTITHERM OVEN 15L 650°C PID
Overige gegevens : Weight : 50.00000kg
Produktcode. 11970098
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »