missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VEGFD Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 26915-1-AP-150UL
This item is not returnable.
View return policy
Beschrijving
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family and is active in angiogenesis, lymphangiogenesis, and endothelial cell growth. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-2 and VEGFR-3 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor C.Specificatie
VEGFD | |
Polyclonal | |
Unconjugated | |
VEGFD | |
c Fos induced growth factor, FIGF, VEGF D, VEGFD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
2277 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43915 | |
VEGFD | |
VEGFD Fusion Protein Ag25552 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction