Learn More
DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls
Clean-processed to maximize cleanliness and have been sterilized by gamma-irradiation. DuPont™ Tyvek™ IsoClean™ Unhooded Coveralls are folded to aid aseptic donning and individually packed in an ISO Class 4 cleanroom.
Merk: DuPont™ D13471957
This item is not returnable.
View return policy
Beschrijving
- Delivers an ideal balance of protection, durability and comfort.
- Made of high density polyethylene using a patented flash spinning process.
- Breathable and exceptionally low linting.
- Garments and accessories made of clean-processed and sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
- Clean-processed and sterilised by gamma-irradiation to SAL of 10-6 (ISO 11137-1)
- Full traceability on all sterilized apparel with certificates of sterility available
- Suitable for use in GMP class A/B (ISO Class 5) clean rooms
- This garment is not CE certified as chemical protective clothing
Specificatie
Cleanroom Coveralls | |
HDPE | |
Elastic | |
Front Zipper | |
Sterile | |
Slip-retardant shoe covers | |
Class 100 (ISO 5) |
Medium | |
White | |
Elastic | |
Bound | |
ISO Class 5 | |
D13471957 |