Learn More
Cole-Parmer Thermoplastic Vulcanizate (TPV) Industrial Tubing
Compatible with most CIP (Clean-in-Place) solutions and SIP (Steam-in-Place)
Merk:  Cole-Parmer 50115-77
 Bekijk meerdere types van dit product via de
Produktcode. 30053817
Dit item is uit de verkoop gehaald en is niet meer verkrijgbaar. Bekijk het product voor mogelijke alternatieven.
Bekijk hier alternatieve producten »
Beschrijving
Beschrijving
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to Santoprene™ tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Specificatie
Specificatie
ADCF;Non-DEHP;Non-Phthalate;RoHS;USP
TPV
50 ft.
EtO with no issues
3/4 in.
1 in.
50 ft.
Tan
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie