Learn More
Cole-Parmer Thermoplastic Vulcanizate (TPV) Industrial Tubing
Compatible with most CIP (Clean-in-Place) solutions and SIP (Steam-in-Place)
Merk: Cole-Parmer 50115-77
This item is not returnable.
View return policy
Beschrijving
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to Santoprene™ tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. USP Class VI, USP 661, and RoHS compliant and BPOG testedSpecificatie
ADCF;Non-DEHP;Non-Phthalate;RoHS;USP | |
TPV | |
50 ft. | |
EtO with no issues |
3/4 in. | |
1 in. | |
50 ft. | |
Tan |