Learn More
Cole-Parmer Thermoplastic Vulcanizate (TPV) Industrial Tubing
Compatible with most CIP (Clean-in-Place) solutions and SIP (Steam-in-Place)
Brand:  Cole-Parmer 50115-78
 View more versions of this product
Product Code. 30053168
  • 86.05 / 15m
Only null left
Description
Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection. E in., cellent for use in peristaltic pump applications. Great alternative to Santoprene™ tubing. Does not contain PVC, DEHP, phthalates, or other plasticizers. Ultra-low e in., tractables and leachables. Low gas and o in., ygen permeability. USP Class VI, USP 661, and RoHS compliant and BPOG tested
Specifications
Specifications
ADCF;Non-DEHP;Non-Phthalate;RoHS;USP
TPV
50 ft.
E-beam/gamma irradiation between 25 and 45 kGy
1/16 in.
3/16 in.
50 ft.
Tan