Learn More
Thermo Scientific™ BetaBasic™ CN Javelin™ Guard Columns
Offer convenient, easy to use protection for analytical HPLC columns
379.00 - 444.00
Specificatie
Partikelgrootte 5 μm
Stationaire fase CN
Meer specificaties weergeven
Producten 2
Produktcode Merk Diameter (metrisch) binnen Lengte (metrisch) Prijs Hoeveelheid en beschikbaarheid  
Produktcode Merk Diameter (metrisch) binnen Lengte (metrisch) Prijs Hoeveelheid en beschikbaarheid  
11583261
Bekijk documenten
Thermo Scientific™
71705-022106
2.1 mm 20 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11593261
Bekijk documenten
Thermo Scientific™
71705-024006
4 mm 20 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Beschrijving
Beschrijving
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Specificatie
Specificatie
5 μm
CN