Learn More
Memmert™ Test Tube Racks with 36 Holes
Test tube rack with 36 holes and 3-row rack
Merk: Memmert™ WTB6B48834
70.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Beschrijving
- Test tube rack
- 36 holes 3-row rack
- Diameter is 18 mm
Specificatie
36 holes, 3- row rack | |
Test Tube Rack | |
18 mm |
WTB6 | |
Transverses for sizes 15 and 24 |