missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TDP-43 Rabbit anti-Human, Mouse, Rat, Zebrafish, kak, Biotin, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech BIOTIN-10782-100UL
This item is not returnable.
View return policy
Beschrijving
TDP-43 Polyclonal antibody apecifically detetects TDP-43 antigen in Human, Mouse, Rat, Zebrafish, kak Samples. It is validated for Immunohistochemistry.Specificatie
TDP-43 | |
Polyclonal | |
Biotin | |
BC001487 | |
Rabbit | |
Antigen affinity purification | |
Primary | |
Human, Mouse, Rat, Zebrafish | |
Liquid |
Immunohistochemistry | |
500 μg/mL | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
TARDBP | |
TDP-43 fusion protein Ag1231 | |
100 μL | |
23435 | |
Store at -20°C. Avoid exposure to light. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction