Learn More
TCR Beta Armenian Hamster anti-Mouse, CoraLite 488, Clone: H57-597, Proteintech
Armenian Hamster Monoclonal Antibody
Merk:  Proteintech CL488-65106-100TESTS
Produktcode. 16870656
  • 124.00 /
Explore more special offers
Beschrijving
Beschrijving
The TCR Beta antibody from Proteintech is a armenian hamster monoclonal antibody generated with affinity purified tcr from mouse do-11.10 cells of mouse TCR Beta. This antibody recognizes mouse antigen. The TCR Beta antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™488.
Specificatie
Specificatie
TCR Beta
Monoclonal
5 μL/Test
Armenian Hamster
Primary
Mouse
Immunofluorescence, Flow Cytometry
H57-597
CoraLite 488
100 Tests
21577
IgG
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen