Learn More
TCR Beta Armenian Hamster anti-Mouse, CoraLite 488, Clone: H57-597, Proteintech
Armenian Hamster Monoclonal Antibody
Merk: Proteintech CL488-65106-100TESTS
Beschrijving
The TCR Beta antibody from Proteintech is a armenian hamster monoclonal antibody generated with affinity purified tcr from mouse do-11.10 cells of mouse TCR Beta. This antibody recognizes mouse antigen. The TCR Beta antibody has been validated for the following applications: IF, FC analysis. This antibody is coralite™488.
Specificatie
TCR Beta | |
Monoclonal | |
5 μL/Test | |
Armenian Hamster | |
Primary | |
Mouse |
Immunofluorescence, Flow Cytometry | |
H57-597 | |
CoraLite 488 | |
100 Tests | |
21577 | |
IgG |