Learn More
Micronova Tape extreme temperature, controlled environment, Polyimide, with silicone adhesive, -70°C to 260°C
Merk:  Micronova 825-15AM
Tape extreme temperature, controlled environment, Polyimide, with silicone adhesive, -70°C to 260°C
Overige gegevens : Weight : 1.00000kg
Produktcode. 12863552
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »