Learn More
Micronova Tape controlled environment, polyethylene, black 38mm x 33m
Merk: Micronova PCX-15BL
Tape controlled environment, polyethylene, black 38mm x 33m
Overige gegevens : Weight : 1.00000kg