Learn More
Micronova Tape conductive grid, controlled environment, 152mm x 33m
Merk:  Micronova CGT-6
Tape conductive grid, controlled environment, 152mm x 33m
Overige gegevens : Weight : 0.20000kg
Produktcode. 12897023
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »