Learn More
Cole-Parmer™ Stuart™ Heating Block Kit
Product Code. 15173027
Click to view available options
Includes:
Base with 1 x 25 mL, 1 x 50 mL and 1 x 100 mL inserts
Base with 3 x 25 mL, 3 x 50 mL and 3 x 100 mL inserts
Base with 3 x 250 mL and 3 x 100 mL inserts
Base with 3 x 250 mL inserts
Base with 4 x 500 mL inserts
Unit Size:
Each
Turn your existing 6 x 6 in. hot plate into a heating mantle
  • Safest way to heat round-bottom flasks
  • Flexible modular system for a wide range of sizes up to 500 mL
  • Low-well design is perfect for multi-neck flasks
Specifications
For Use With (Equipment) Hot Plate or Stirring Hot Plate
Material Anodized Aluminum
Type Heating Block
Includes Base with 3 x 250 mL and 3 x 100 mL inserts