Learn More
STAT3 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk:  Proteintech CL488-10253-100UL
Produktcode. 17442463
  • 450.00 /
Beschrijving
Beschrijving
The STAT3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human STAT3. This antibody recognizes human, mouse, rat antigen. The STAT3 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specificatie
Specificatie
STAT3
Polyclonal
Rabbit
Primary
Human, Rat, Mouse
Flow Cytometry (Intracellular Staining)
CoraLite 488
100 μL
6774