Learn More
STAT3 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech CL488-10253-100UL
This item is not returnable.
View return policy
Beschrijving
The STAT3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human STAT3. This antibody recognizes human, mouse, rat antigen. The STAT3 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™488.
Specificatie
STAT3 | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Rat, Mouse |
Flow Cytometry (Intracellular Staining) | |
CoraLite 488 | |
100 μL | |
6774 | |
IgG |