Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN750, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Merk:  Memmert™ SN750B02075
Produktcode. 17598715
  • 298.00 /
Explore more special offers
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specificatie
Specificatie
SN750 Sterilizer
Drip Tray
Rim: 15 mm
Stainless Steel
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie
Certificates
Speciale aanbiedingen
Speciale aanbiedingen