Learn More
Memmert™ Stainless Steel Slide-In Bottom Drip Tray for SN160, 15 mm
Non-perforated slide-in drip trays constructed of stainless steel.
Merk:  Memmert™ SN160B02073
133.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Produktcode. 17568495
  • 139.00 / Each
Only null left
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Accessory
  • May affect temperature distribution
  • Constructed of stainless steel
  • Rim height: 15 mm
Specificatie
Specificatie
SN160 Sterilizer
Drip Tray
Rim: 15 mm