Learn More
Scientific Industries SI™ Shock absorbing feet for Vortex-Genie™
Upgrade the standard feet of your Vortex Genie™ with shock-absorbing Sorbothane™ rubber feet. Scientific Industries SI™ Shock Absorbing Feet for Vortex-Genie™ come with screws for easy mounting with a standard screwdriver.
Merk: Scientific Industries SI™ 0K-0236-900
This item is not returnable.
View return policy
Beschrijving
- Fits on all Vortex-Genie™ mixers
- Fits on Disruptor Genie™
- Includes rubber feet and four mounting screws
- Easily mounted with a standard screwdriver
- If your bottom closure is made of black plastic, you cannot use these replacement feet. Please order P/N OK-0236-408.
Specificatie
Shock-absorbing rubber feet | |
Sorbothane™ rubber | |
Vortex-Genie™ mixers and Disruptor Genie™ | |
4/Pk. |