Learn More
Ansell™ HyFlex™ 48-105 Series White Polyurethane Light Weight Gloves
Designed for light applications that require good levels of comfort and abrasion resistance.
Merk:  Ansell™ 48-105_9
 Bekijk meerdere types van dit product via de
Produktcode. 13458958
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Beschrijving
Beschrijving
  • Industrial gloves
  • Comfort and protection for lightweight applications
  • Offers precise handling of small objects in dry or light oily environments
  • Provides enhanced safety, dexterity and durability
Applications:
  • Assembling, inspecting and packing small components
  • Light assembly tasks
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Polyurethane
Nylon
9
Knit
12 Pairs
48-105
Standard CATEGORY II EN 388 : 0030
Lightweight Gloves
White
Nylon
Abrasion resistance