Learn More
Brady™ Self-laminating White Vinyl Wrap Around Wire and Cable Label Tape for M6/M7 Printers
Self-laminating vinyl material is ideal for wire and cable identification. Designed with a clear protective layer to cover the printed text, B-427 material provides superb protection from abrasion, dirt, oils, solvents, water and excess handling.
29.95 - 142.00
Specificatie
Autoclaveerbaar | Not Autoclavable |
---|---|
Certificeringen/conformiteit | ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized |
Type kleefmiddel | Permanent Acrylic |
Coating | None |
Kleur | Clear, White |
Produktcode | Merk | Dimensions (LxW) | Voor gebruik met (apparatuur) | Indeling | Hoogte (metrisch) | Prijs | Hoeveelheid en beschikbaarheid | ||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Merk | Dimensions (LxW) | Voor gebruik met (apparatuur) | Indeling | Hoogte (metrisch) | Prijs | Hoeveelheid en beschikbaarheid | ||
17821589
|
Brady™
173691 |
0.75 x 0.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 19.05 mm |
52.05
Pack of 750 |
|
||
17879194
|
Brady™
174532 |
0.75 x 0.25 in. | M710, BMP71 | Die-cut | 19.05 mm |
54.35
Pack of 750 |
|
||
17878724
|
Brady™
173429 |
0.75 x 0.5 in. | M710, BMP71 | Die-cut | 19.05 mm |
58.10
Pack of 500 |
|
||
17878274
|
Brady™
174180 |
0.75 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 19.05 mm |
62.75
Pack of 500 |
|
||
17878874
|
Brady™
174660 |
0.75 x 1 in. | M710, BMP71 | Die-cut | 19.05 mm |
46.85
Pack of 250 |
|
||
17891689
|
Brady™
173917 |
0.75 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 19.05 mm |
44.45
Pack of 250 |
|
||
17818794
|
Brady™
173403 |
1 x 0.75 in. | M710, BMP71 | Die-cut | 25.4 mm |
50.60
Pack of 250 |
|
||
17898194
|
Brady™
174216 |
1 x 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
54.70
Pack of 250 |
|
||
17829144
|
Brady™
173353 |
1.5 x 0.75 in. | M710, BMP71 | Die-cut | 38.1 mm |
56.60
Pack of 250 |
|
||
17809204
|
Brady™
174267 |
1.5 x 0.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
61.15
Pack of 250 |
|
||
17838474
|
Brady™
174208 |
1 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
66.85
Pack of 500 |
|
||
17889044
|
Brady™
173411 |
1 x 0.5 in. | M710, BMP71 | Die-cut | 25.4 mm |
61.90
Pack of 500 |
|
||
17818904
|
Brady™
173399 |
1 x 1 in. | M710, BMP71 | Die-cut | 25.4 mm |
55.60
Pack of 250 |
|
||
17878464
|
Brady™
174221 |
1 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
75.05
Pack of 250 |
|
||
17821389
|
Brady™
173694 |
1 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
85.70
Pack of 250 |
|
||
17898744
|
Brady™
174533 |
1 x 1.5 in. | M710, BMP71 | Die-cut | 25.4 mm |
89.50
Pack of 250 |
|
||
17801669
|
Brady™
173914 |
1 x 1.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 25.4 mm |
141.00
Pack of 250 |
|
||
17848784
|
Brady™
174593 |
1.5 x 0.25 in. | M710, BMP71 | Die-cut | 38.1 mm |
69.60
Pack of 750 |
|
||
17828864
|
Brady™
173362 |
1.5 x 0.5 in. | M710, BMP71 | Die-cut | 38.1 mm |
75.75
Pack of 500 |
|
||
17899014
|
Brady™
173345 |
1.5 x 1 in. | M710, BMP71 | Die-cut | 38.1 mm |
71.75
Pack of 250 |
|
||
17888894
|
Brady™
173337 |
1.5 x 1.5 in. | M710, BMP71 | Die-cut | 38.1 mm |
73.35
Pack of 250 |
|
||
17878754
|
Brady™
174651 |
1.5 x 1.75 in. | M710, BMP71 | Die-cut | 38.1 mm |
85.70
Pack of 250 |
|
||
17889194
|
Brady™
174534 |
1.5 x 2 in. | M710, BMP71 | Die-cut | 38.1 mm |
94.50
Pack of 100 |
|
||
17811699
|
Brady™
173915 |
1.5 x 1.75 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
82.15
Pack of 250 |
|
||
17828734
|
Brady™
174531 |
1.25 x 1 in. | M710, BMP71 | Die-cut | 31.75 mm |
61.90
Pack of 250 |
|
||
17868334
|
Brady™
174275 |
1.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
77.40
Pack of 250 |
|
||
17838404
|
Brady™
174283 |
1.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
79.30
Pack of 250 |
|
||
17851519
|
Brady™
173780 |
1.5 x 0.25 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
66.65
Pack of 750 |
|
||
17818404
|
Brady™
174257 |
1.5 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
81.90
Pack of 500 |
|
||
17841609
|
Brady™
173688 |
1.25 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 31.75 mm |
35.05
Pack of 250 |
|
||
17841579
|
Brady™
173689 |
1.25 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 31.75 mm |
59.35
Pack of 250 |
|
||
17831539
|
Brady™
173700 |
1.5 x 2 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 38.1 mm |
90.55
Pack of 100 |
|
||
17841589
|
Brady™
173899 |
2.2 x 0.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 55.88 mm |
29.95
Pack of 100 |
|
||
17881559
|
Brady™
173900 |
2.625 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 66.68 mm |
51.90
Pack of 100 |
|
||
17808304
|
Brady™
174236 |
2.5 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 63.5 mm |
57.55
Pack of 100 |
|
||
17881399
|
Brady™
173716 |
2.5 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 63.5 mm |
51.20
Pack of 100 |
|
||
17889204
|
Brady™
173378 |
2.5 x 1 in. | M710, BMP71 | Die-cut | 63.5 mm |
42.70
Pack of 100 |
|
||
17829164
|
Brady™
174640 |
3.15 x 1.938 in. | M710, BMP71 | Die-cut | 80 mm |
72.30
Pack of 100 |
|
||
17818224
|
Brady™
173316 |
3.375 x 1 in. | M710, BMP71 | Die-cut | 85.73 mm |
47.85
Pack of 100 |
|
||
17818444
|
Brady™
173894 |
3.15 x 1.938 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 80.01 mm |
78.10
Pack of 100 |
|
||
17838304
|
Brady™
174338 |
3.375 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 85.73 mm |
39.55
Pack of 100 |
|
||
17801679
|
Brady™
173703 |
4 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 101.6 mm |
57.80
Pack of 100 |
|
||
17868214
|
Brady™
174243 |
4 x 1 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 101.6 mm |
54.95
Pack of 100 |
|
||
17848124
|
Brady™
174285 |
4 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 101.6 mm |
74.40
Pack of 100 |
|
||
17819024
|
Brady™
174535 |
4 x 1.5 in. | M710, BMP71 | Die-cut | 101.6 mm |
60.35
Pack of 100 |
|
||
17858894
|
Brady™
173335 |
4 x 1.5 in. | M710, BMP71 | Die-cut | 101.6 mm |
68.85
Pack of 100 |
|
||
17858984
|
Brady™
173372 |
4 x 1 in. | M710, BMP71 | Die-cut | 101.6 mm |
50.85
Pack of 100 |
|
||
17829174
|
Brady™
173332 |
6 x 1.5 in. | M710, BMP71 | Die-cut | 152.4 mm |
57.05
Pack of 50 |
|
||
17858214
|
Brady™
174288 |
6 x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Die-cut | 152.4 mm |
61.70
Pack of 50 |
|
||
17861629
|
Brady™
173696 |
50 ft. x 1.5 in. | M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter) | Continuous | 38.1 mm |
142.00
1 roll |
|
||
Beschrijving
- Provided in a self-laminated format with a white printable zone and translucent overlaminating area to protect text
- Durable, permanent acrylic adhesive has outstanding water and oil resistance
- UL approved
- Service temperature of -40°C to +70°C (-40°F to +158°F)
- Made of B-427 vinyl material
Specificatie
Not Autoclavable | |
Permanent Acrylic | |
Clear, White | |
Vinyl | |
Thermal Transfer | |
Non-sterile | |
No | |
-70°C to +70°C | |
0.102 mm |
ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized | |
None | |
Matte | |
Single-side Printable | |
Labels | |
Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling | |
-94°F to +158°F | |
0.004 in. |