Learn More
Ansell™ Scorpio™ 8-352 Series Green Neoprene Immersion Gloves
Neoprene dipped coating on interlock knit liner for better grip on wet and slippery materials.
Merk:  Ansell™ 08-352 10
 Bekijk meerdere types van dit product via de
Produktcode. 10452204
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Bekijk hier alternatieve producten »
Beschrijving
Beschrijving
  • Flexible
  • Textured finish
  • 100% liquidproof
  • Soft stretchy knit liner with no seams in working area
  • Comfortable to wear, cushions the hand, absorbs perspiration and prevents chafing
  • Good wet grip. Safer handling means increased productivity
  • Easier to manipulate, less tiring to the hand and easier to get on and off
  • Neoprene coating
  • Wide ranging chemical protection
  • Certified FDA21 CFR 177-2600
Applications:
  • Cleaning furnaces, distillation pumps, valves or lines and crackers BTX process
  • Equipment repair and maintenance
  • Filling, blending and charging of raw materials
  • Loading and unloading process equipment
  • Opening and draining pumps valves or lines
  • Opening furnaces draining pumps valves or lines and crackers BTX process
  • Transferring liquids and solids
  • Transferring liquids and solids between vessels and tanks and process equipment
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Neoprene
Neoprene
Yes
8-352
Interlock Knit Cotton
72 Pairs
8-352
ANSI, EN 388 : 3121, EN 374
Neoprene
Immersion Gloves
Gauntlet
Fully coated-palm
Chemicals
Product Suggestions
Product Suggestions
Video's
SDS
Documentatie
Documentatie