missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SCARF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech 13702-1-AP-20UL
This item is not returnable.
View return policy
Beschrijving
The protein encoded by this gene is a scavenger receptor that is expressed in endothelial cells and mediates the binding and degradation of acetylated low density lipoprotein (Ac-LDL). Transcription factors ZNF444/EZF-2 and SP1 are found to be responsible for the regulation of this gene. Five alternatively spliced transcript variants encoding distinct isoforms have been described.Specificatie
SCARF1 | |
Polyclonal | |
Unconjugated | |
SCARF1 | |
Acetyl LDL receptor, KIAA0149, SCARF1, SREC, SREC-I | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303313, 380713, 8578 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q14162, Q5ND28 | |
Scarf1 | |
SCARF1 Fusion Protein Ag4621 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction