Learn More
Sartorius Interface Module for Cubis Balances
GREENER_CHOICE
Interface Module for use with Cubis balances.
Merk:  Sartorius YDO01MS-B
220.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 Bekijk meerdere types van dit product via de
Produktcode. 13478038
  • 228.00 / 1 stuk
Beschrijving
Beschrijving
  • Option R (YDO01MS-R): RS-232C data interface, 25-pin, for connection of Cubis accessory equipment
  • Option B (YDO01MS-B): Bluetooth™ interface module for wireless connection of the YDP10BT data printer
  • Option P (YDO01MS-P): RS-232C data interface, 9-pin, including PS/2 port for connecting a PC or a keyboard
TRUSTED_SUSTAINABILITY
Specificatie
Specificatie
Cubis Balance
Option B: Bluetooth Adapter
Wireless connection of the YDP10BT data printer
Data Interface