Learn More
S100A11 Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Merk: Proteintech CL488-10237-100UL
This item is not returnable.
View return policy
Beschrijving
The S100A11 antibody from Proteintech is a rabbit polyclonal antibody to a recombinant protein of human S100A11. This antibody recognizes human, rat, mouse antigen. The S100A11 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™488.
Specificatie
S100A11 | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Mouse, Rat |
Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
CoraLite 488 | |
100 μL | |
6282 | |
IgG |